Thallium(I) hexafluoroacetylacetonate ≥99% (99.9%-Tl)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Thallium(I) hexafluoro-2,4-pentanedionate
81-2500-1G
81-2500-5G
100210-100EA
62.28
USD
100210-100
100196-404
Thallium(I) hexafluoroacetylacetonate ≥99% (99.9%-Tl)
Thallium (I) hexafluoroacetylacetonate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₅H₃F₁₈O₆Tl Boiling Pt: 220 °C/ 140 °C (0.1 mmHg) (dec./ subl.) Melting Pt: 126…128 °C |
MDL Number:
MFCD00064761 CAS Number: 15444-43-6 UN: 1707 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...