Trimethyl(methylcyclopentadienyl)platinum(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trimethyl (methylcyclopentadienyl) platinium (IV), Platinum, trimethyl(2-methyl-1,3-cyclopentadien-1-yl)-, TRIMETHYL(METHYLCYCLOPENTADIENYL)PLATINUM

78-1350-2G 78-1350-500MG
100208-682EA 955.38 USD
100208-682 100196-380
Trimethyl(methylcyclopentadienyl)platinum(IV) ≥99%
Trimethyl(methylcyclopentadienyl)platinum(IV)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₉H₁₆Pt
Boiling Pt: 23 °C (0.053 mmHg) (subl.)
Melting Pt: 30…31 °C
Density: 1,88
MDL Number: MFCD00079665
CAS Number: 94442-22-5
UN: 3467
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR