Barium tetracyanoplatinate(II) tetrahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

78-0083-5G 78-0083-1G
100208-060EA 1314.89 USD
100208-060 100196-370
Barium tetracyanoplatinate(II) tetrahydrate ≥99%
Barium tetracyanoplatinate(II) tetrahydrate
Formula: BaPt(CN)₄.4H₂O
Density: 2,076
MDL Number: MFCD00014183
CAS Number: 13755-32-3
UN: 3288
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR