Sodium hexachloroiridate(III) hydrate

Supplier: STREM CHEMICALS INC MS
Warning

77-9000-1G 77-9000-5G
100196-364EA 250.52 USD
100196-364 100210-094
Sodium hexachloroiridate(III) hydrate
Sodium hexachloroiridate(III) hydrate
Formula: Na₃IrCl₆·xH₂O
MDL Number: MFCD00003508
CAS Number: 123334-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR