Tetrabutylammonium tetrathiorhenate(VII) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

75-1000-5G 75-1000-1G
100206-800EA 639.75 USD
100206-800 100206-798
Tetrabutylammonium tetrathiorhenate(VII) ≥99%
Tetrabutylammonium tetrathiorhenate(VII)
MDL Number: MFCD00145366
CAS Number: 16829-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR