Dibutylbis(2,4-pentanedionato-O,O')tin
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Di-n-butyltinbis(acetylacetonate), Di-n-butyltin bis(2,4-pentanedionate), (OC-6-11)-Dibutylbis(2,4-pentanedionato-κO2,κO4)Tin
50-1050-100G
50-1050-500G
100211-136EA
56.62
USD
100211-136
100200-284
Dibutylbis(2,4-pentanedionato-O,O')tin
Dibutylbis(2,4-pentanedionato-O,O')tin
Formula:
C₁₈H₃₂O₄Sn Boiling Pt: 150 °C (3 mmHg) Melting Pt: 26 °C Density: 1,21 |
MDL Number:
MFCD00077994 CAS Number: 22673-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...