Trimethylindium ≥98% (99.9+%-In)

Supplier: STREM CHEMICALS INC MS
Danger

49-2010-10G 49-2010-2G
100211-132EA 1186.09 USD
100211-132 100211-134
Trimethylindium ≥98% (99.9+%-In)
Trimethylindium

Metal Alkyls, Volatile Precursors for CVD

Melting Pt: 88 °C
Density: 1,568
Flash Pt: –1 °F
MDL Number: MFCD00014846
CAS Number: 3385-78-2
UN: 3393
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR