Tris(dibenzylideneacetone)dipalladium(0)

Supplier: STREM CHEMICALS INC MS

Synonyms: Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]

46-3000-5G 46-3000-1G
100198-026EA 389.23 USD
100198-026 100201-686
Tris(dibenzylideneacetone)dipalladium(0)
Tris(dibenzylideneacetone)dipalladium(0)
Formula: C₅₁H₄₂O₃Pd₂
Melting Pt: 152…155 °C
MDL Number: MFCD00013310
CAS Number: 51364-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR