Palladium 5% on calcium carbonate, unpoisoned, reduced

Supplier: STREM CHEMICALS INC MS

46-2010-10G 46-2010-50G
100198-208EA 307.14 USD
100198-208 100198-210
Palladium 5% on calcium carbonate, unpoisoned, reduced
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR