Palladium 5% on barium carbonate, reduced
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-1970-10G
46-1970-50G
100204-072EA
199.57
USD
100204-072
100210-650
Palladium 5% on barium carbonate, reduced
Palladium
Formula:
Pd |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...