Palladium 10% on activated wood carbon wetted with ca. 50% water, unreduced (Escat™ 1921)

Supplier: STREM CHEMICALS INC MS
Warning

46-1906-10G 46-1906-50G
100200-990EA 747.32 USD
100200-990 100200-992
Palladium 10% on activated wood carbon wetted with ca. 50% water, unreduced (Escat™ 1921)
Palladium

Escat

Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR