Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer

45-0380-250MG 45-0380-5G 45-0380-1G
100196-244EA 461.42 USD
100196-244 100203-172 100208-672
Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98%
Chloro(1,5-cyclooctadiene)rhodium (I) dimer
Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
MDL Number: MFCD00012415
CAS Number: 12092-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR