Potassium perruthenate(VII) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Potassium perruthenate

44-1750-5G 44-1750-1G
100206-654EA 554.83 USD
100206-654 100206-652
Potassium perruthenate(VII) ≥98%
Potassium perruthenate(VII)
Formula: C₃₂H₃₆N₂O₅
MDL Number: MFCD00061333
CAS Number: 10378-50-4
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR