Potassium hexacyanoruthenate(II) hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium hexacyanoruthenate hydrate

44-1700-5G 44-1700-1G 44-1700-250MG
100205-950EA 1043.14 USD
100205-950 100210-648 100200-470
Potassium hexacyanoruthenate(II) hydrate
Potassium hexacyanoruthenate(II) hydrate
Formula: C₆K₄N₆Ru·xH₂O
MDL Number: MFCD00150398
CAS Number: 339268-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR