Hexaammineruthenium trichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

44-0620-1G 44-0620-250MG 44-0620-5G
100198-190EA 165.6 USD
100198-190 100201-646 100200-980
Hexaammineruthenium trichloride ≥99%
Hexaammineruthenium trichloride

Metallocenes, Derivatives & Cp Precursors

Formula: H₁₈Cl₃N₆Ru
MDL Number: MFCD00011478
CAS Number: 14282-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR