[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dichloro[(S)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II), Dichloro [(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
44-0249-500MG
44-0249-2G
100199-910EA
236.37
USD
100199-910
100209-346
[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride ≥95%
[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride
BINAP
Formula:
C₄₄H₃₄Cl₂P₂Ru |
MDL Number:
MFCD01073794 CAS Number: 134524-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...