Strontium neodecanoate (16 - 21% Sr), Superconductor Grade

Supplier: STREM CHEMICALS INC MS

38-7000-50G 38-7000-10G
100205-106EA 553.42 USD
100205-106 100208-024
Strontium neodecanoate (16 - 21% Sr), Superconductor Grade
Strontium neodecanoate
Formula: Sr[OOCC(CH₃)₂(CH₂)₅CH₃]₂
MDL Number: MFCD00070439
CAS Number: 106705-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR