Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')gallium(III) ≥99% (99.999%-Ga), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III), Ga(TMHD)3

31-5000-1G 31-5000-5G 31-5000-25G
100201-594EA 133.05 USD
100201-594 100205-928 100207-382
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')gallium(III) ≥99% (99.999%-Ga), PURATREM
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')gallium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇GaO₆
Boiling Pt: 360 °C/ 170 °C (0.2 mmHg) (dec./ subl.)
Melting Pt: 219…220 °C
MDL Number: MFCD00058705
CAS Number: 34228-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR