Zinc protoporphyrin

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX

30-3090-10MG 30-3090-50MG
100196-196EA 67.94 USD
100196-196 100201-592
Zinc protoporphyrin
Zinc protoporphyrin
Formula: C₃₄H₃₂N₄O₄Zn
MDL Number: MFCD00011612
CAS Number: 15442-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR