Nickel skeletal, unpromoted (Actimet® M)

Supplier: STREM CHEMICALS INC MS
Danger


Supplied under water.

28-1916-250G 28-1916-50G
100200-672EA 1460.68 USD
100200-672 100201-574
Nickel skeletal, unpromoted (Actimet® M)
Nickel
Formula: Ni
MDL Number: MFCD00011137
CAS Number: 7440-02-0
UN: 1378
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR