Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) ≥99% (99.9+%-Co)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), Co(TMHD)3
27-3000-1G
27-3000-5G
27-3000-25G
100200-670EA
131.63
USD
100200-670
100210-804
100203-762
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) ≥99% (99.9+%-Co)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇CoO₆ Boiling Pt: 250 °C/ 120 °C (0,5 mmHg) (dec./ subl.) Melting Pt: 254…256 °C |
MDL Number:
MFCD00233617 CAS Number: 14877-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...