Ferrocenecarbaldehyde ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Ferrocenecarboxaldehyde

26-1750-50G 26-1750-2G 26-1750-10G
100202-762EA 638.34 USD
100202-762 100206-508 100208-866
Ferrocenecarbaldehyde ≥98%
Ferrocenecarbaldehyde
Formula: C₁₁H₁₀FeO
Melting Pt: 124 °C
MDL Number: MFCD00001429
CAS Number: 12093-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR