Ferroceneacetic acid ≥98%

Supplier: STREM CHEMICALS INC MS

26-1430-2G 26-1430-500MG
100198-726EA 544.92 USD
100198-726 100209-854
Ferroceneacetic acid ≥98%
Ferroceneacetic acid
Formula: C₁₂H₁₂FeO₂
Melting Pt: 158…160 °C
MDL Number: MFCD00045464
CAS Number: 1287-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR