N,N-Dimethylaminomethylferrocene ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferrocenylmethyldimethylamine, (Dimethylaminomethyl)ferrocene

26-1400-5G 26-1400-100G 26-1400-25G
100207-626EA 59.45 USD
100207-626 100196-926 100200-908
N,N-Dimethylaminomethylferrocene ≥95%
N,N-Dimethylaminomethylferrocene

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₃H₁₇FeN
Boiling Pt: 91…92 °C (0.5 mmHg)
Density: 1,228
Flash Pt: >230 °F
MDL Number: MFCD00001433
CAS Number: 1271-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR