Methylcyclopentadienylmanganese tricarbonyl ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

25-1550-5G 25-1550-25G
100207-624EA 111.82 USD
100207-624 100207-622
Methylcyclopentadienylmanganese tricarbonyl ≥97%
Methylcyclopentadienylmanganese tricarbonyl

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₉H₇MnO₃
Boiling Pt: 233 °C
Melting Pt: –2.2 °C
Density: 1.38
Flash Pt: 205 °F
MDL Number: MFCD00001426
CAS Number: 12108-13-3
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR