Vanadium naphthenate oxide 35% in naphthenic acid (2.8-3.2% V)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Vanadyl naphthenate
23-4400-500G
23-4400-100G
23-4400-2500G
100208-854EA
593.05
USD
100208-854
100209-784
100200-892
Vanadium naphthenate oxide 35% in naphthenic acid (2.8-3.2% V)
Vanadium naphthenate oxide
Formula:
C₄₄H₂₈O₈V Density: 1.04 Flash Pt: >350 °F |
MDL Number:
MFCD00148763 CAS Number: 68553-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...