Tetrachlorobis(tetrahydrofuran)titanium(IV) ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

22-1080-5G 22-1080-25G 22-1080-100G
100196-146EA 92 USD
100196-146 100209-778 100198-702
Tetrachlorobis(tetrahydrofuran)titanium(IV) ≥97%
Tetrachlorobis(tetrahydrofuran)titanium(IV)
Formula: C₄H₈Cl₄OTi
Melting Pt: 126…128 °C
MDL Number: MFCD00077884
CAS Number: 31011-57-1
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR