Chlorotitanium triisopropoxide ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
22-0380-100G
22-0380-25G
100200-650EA
322.71
USD
100200-650
100200-652
Chlorotitanium triisopropoxide ≥97%
Chlorotitanium triisopropoxide
Alkoxide
Formula:
C₉H₂₁ClO₃Ti Melting Pt: 35…40 °C Density: 1.091 Flash Pt: 72 °F |
MDL Number:
MFCD00009861 CAS Number: 20717-86-6 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...