Potassium tris(1-pyrazolyl)borohydride hydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Potassium hydrotris(pyrazol-1-yl)borate hydrate, Potassium hydrotris(1-pyrazolyl)borate hydrate
19-3600-5G
19-3600-1G
100197-436EA
527.94
USD
100197-436
100197-434
Potassium tris(1-pyrazolyl)borohydride hydrate ≥98%
Potassium tris(1-pyrazolyl)borohydride hydrate
Formula:
C₉H₁₀BKN₆·xH₂O Melting Pt: 185…189 °C |
MDL Number:
MFCD03414149 CAS Number: 18583-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...