Tricyclohexylphosphonium tetrafluoroborate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-6160-1G 15-6160-5G
100206-410EA 42.46 USD
100206-410 100206-412
Tricyclohexylphosphonium tetrafluoroborate ≥99%
Tricyclohexylphosphonium tetrafluoroborate
Formula: C₁₈H₃₄BF₄P
MDL Number: MFCD03840580
CAS Number: 58656-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR