Tricyclohexylphosphine 20% in toluene ≥88%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
15-6151-500G
15-6151-100G
100196-104EA
438.77
USD
100196-104
100197-424
Tricyclohexylphosphine 20% in toluene ≥88%
Tricyclohexylphosphine
Phosphine
Formula:
C₁₈H₃₃P Density: 0.9 Flash Pt: 40 °F |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...