(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-phenoxy ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-ShiP

15-5157-500MG 15-5157-100MG
101418-680EA 174.09 USD
101418-680 101418-682
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-phenoxy ≥98%
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-phenoxy

MONOPHOS, Phosphine

Melting Pt: 102…103 °C
MDL Number: MFCD08459341
CAS Number: 885701-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR