Phosphonitrilic chloride trimer ≥98.5%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene

15-4530-50G 15-4530-10G
100198-658EA 250.52 USD
100198-658 100209-730
Phosphonitrilic chloride trimer ≥98.5%
Phosphonitrilic chloride trimer

Phosphine

Formula: Cl₆N₃P₃
Boiling Pt: 127 °C (12 mmHg)
Melting Pt: 128.8 °C
Density: 1,98
MDL Number: MFCD00006474
CAS Number: 940-71-6
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR