Octylphosphonic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid

15-3520-5G 15-3520-1G
101442-430EA 300.06 USD
101442-430 101418-662
Octylphosphonic acid ≥97%
Octylphosphonic acid
Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Melting Pt: 102…103 °C
MDL Number: MFCD00015841
CAS Number: 4724-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR