(S)-(+)-Neomenthyldiphenylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-3490-500MG 15-3490-2G
100206-388EA 294.4 USD
100206-388 100210-728
(S)-(+)-Neomenthyldiphenylphosphine ≥98%
(S)-(+)-Neomenthyldiphenylphosphine

Phosphine

Formula: C₂₂H₂₉P
Melting Pt: 90…92 °C
MDL Number: MFCD00013415
CAS Number: 43077-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR