(-)-6,6'-{[(1S,3S)-1,3-Dimethyl-1,3-propanediyl]bis(oxy)}bis[4,8-bis(t-butyl)-2,10-dimethoxy-bibenzo[d,f][1,3,2]dioxaphosphepin] ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-Chiraphite

15-1456-500MG 15-1456-100MG
101324-612EA 597.29 USD
101324-612 100198-638
(-)-6,6'-{[(1S,3S)-1,3-Dimethyl-1,3-propanediyl]bis(oxy)}bis[4,8-bis(t-butyl)-2,10-dimethoxy-bibenzo[d,f][1,3,2]dioxaphosphepin] ≥95%
(-)-6,6'-{[(1S,3S)-1,3-Dimethyl-1,3-propanediyl]bis(oxy)}bis[4,8-bis(t-butyl)-2,10-dimethoxy-bibenzo[d,f][1,3,2]dioxaphosphepin]

Phosphine

Formula: C₄₉H₆₆O₁₀P₂
MDL Number: MFCD06798297
CAS Number: 852042-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR