(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

15-1242-1G 15-1242-5G
100202-986EA 70.77 USD
100202-986 100202-988
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

Phosphine, Pincer Ligands and Complexes, XANTPHOS

Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Melting Pt: 221…222 °C
MDL Number: MFCD00233866
CAS Number: 161265-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR