Cyclohexylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

15-0950-50G 15-0950-10G
100206-332EA 607.2 USD
100206-332 100206-330
Cyclohexylphosphine ≥97%
Cyclohexylphosphine

Phosphine

Boiling Pt: 145 °C
Density: 0,8750
MDL Number: MFCD00014297
CAS Number: 822-68-4
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR