Cyclohexyldiphenylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Diphenylcyclohexylphosphine

15-0900-5G 15-0900-1G
100207-344EA 246.28 USD
100207-344 100197-398
Cyclohexyldiphenylphosphine ≥98%
Cyclohexyldiphenylphosphine

Phosphine

Formula: C₁₈H₂₁P
MW: 268.33 g/mol
Melting Pt: 58…62 °C
MDL Number: MFCD00046360
CAS Number: 6372-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR