tert-Butyldichlorophosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
t-Butyldichlorophosphine
15-0500-10G
15-0500-2G
100204-938EA
142.95
USD
100204-938
100201-458
tert-Butyldichlorophosphine ≥98%
tert-Butyldichlorophosphine
Phosphine
Formula:
C₄H₉Cl₂P Boiling Pt: 146…148 °C Melting Pt: 47…48 °C |
MDL Number:
MFCD00013615 CAS Number: 25979-07-1 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...