(S)-(-)-2,2'-Bis[di(3,4,5-trimethoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0159-100MG 15-0159-500MG
101418-550EA 140.12 USD
101418-550 101418-552
(S)-(-)-2,2'-Bis[di(3,4,5-trimethoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(S)-(-)-2,2'-Bis[di(3,4,5-trimethoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

MDL Number: MFCD09753007
CAS Number: 256235-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR