Polysiloxane thio-functionalised metal scavenger (Deloxan® MP)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Deloxan® (Thio-Functionalized Polysiloxane)
14-1871-50G
14-1871-10G
101442-338EA
653.91
USD
101442-338
101442-336
Polysiloxane thio-functionalised metal scavenger (Deloxan® MP)
Polysiloxane thio-functionalised
Deloxan® MP Metal Scavengers (Thio-Functionalized Polysiloxane)
Color and Form: Abrasion-proof spheres; Water content: 50-70%
Physical Characteristics: surface area 300-450m2/g (BET), pore volume 2.5-3.5 ml/g, density 0.55-0.65 kg/L (wet form)
Sold in collaboration with Evonik for research purposes only.