Magnesium ethanolate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Magnesium ethoxide, Magnesium ethylate
12-1250-500G
12-1250-100G
100202-750EA
261.85
USD
100202-750
100206-290
Magnesium ethanolate ≥98%
Magnesium ethanolate
Alkoxide
Formula:
C₄H₁₀MgO₂ MW: 114.43 g/mol Melting Pt: Dec. |
MDL Number:
MFCD00009060 CAS Number: 2414-98-4 UN: 3178 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...