2,2,6,6-Tetramethyl-3,5-octanedione ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: (TMOD)

08-2100-5G 08-2100-1G
100198-548EA 403.38 USD
100198-548 100198-546
2,2,6,6-Tetramethyl-3,5-octanedione ≥98%
2,2,6,6-Tetramethyl-3,5-octanedione
MDL Number: MFCD02684546
CAS Number: 78579-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR