(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
08-0620-1G
08-0620-250MG
100198-516EA
322.71
USD
100198-516
100209-564
(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98%
(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
Formula:
C₂₄H₂₀Br₂O₄ Melting Pt: 133 °C |
MDL Number:
MFCD03788936 CAS Number: 179866-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...