1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,4,8,11-Tetramethylcyclam

07-2112-1G 07-2112-5G
100209-554EA 234.95 USD
100209-554 100209-556
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥98%
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
Formula: C₁₄H₃₂N₄
Melting Pt: 34…36 °C
Flash Pt: >230 °F
MDL Number: MFCD00005106
CAS Number: 41203-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR