N-Benzyl-1,4,8,11-tetraazacyclotetradecane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Benzyl-1,4,8,11-tetraazacyclotetradecane

07-1957-100MG 07-1957-500MG
101442-402EA 259.02 USD
101442-402 101442-404
N-Benzyl-1,4,8,11-tetraazacyclotetradecane ≥98%
N-Benzyl-1,4,8,11-tetraazacyclotetradecane
Formula: C₁₇H₃₀N₄
MDL Number: MFCD09263311
CAS Number: 132723-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR