2,2':6',2''-Terpyridine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
TERPY
07-1920-5G
07-1920-1G
100200-628EA
675.14
USD
100200-628
100202-484
2,2':6',2''-Terpyridine ≥98%
2,2':6',2''-Terpyridine
Pincer Ligands and Complexes
Formula:
C₁₅H₁₁N₃ MW: 233.27 g/mol Melting Pt: 88…89 °C |
MDL Number:
MFCD00006213 CAS Number: 1148-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...