Poly(4-vinylpyridine) (cross-linked), anhydrous, powder, (toluene conditioned)

Supplier: STREM CHEMICALS INC MS

Synonyms: Poly-4-vinylpyridine

07-1770-25G 07-1770-100G 07-1770-500G
100198-076EA 339.69 USD
100198-076 100201-348 100202-612
Poly(4-vinylpyridine) (cross-linked), anhydrous, powder, (toluene conditioned)
Poly(4-vinylpyridine) (cross-linked)
Formula: [-CH₂CH(C₅H₄N)CH₂-]n
Boiling Pt: 225 °C/ 260 °C (dec. air/ dec. N₂)
Density: 1.15
MDL Number: MFCD00190588
CAS Number: 9017-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR