Poly(4-vinylpyridine) (cross-linked) (50-60% H2O), beads
Supplier: STREM CHEMICALS INC MS
Synonyms:
Poly-4-vinylpyridine
07-1755-50G
07-1755-250G
07-1755-1000G
100207-934EA
148.62
USD
100207-934
100207-932
100208-760
Poly(4-vinylpyridine) (cross-linked) (50-60% H2O), beads
Poly(4-vinylpyridine) (cross-linked)
Ionic Liquids
Formula:
[-CH₂CH(C₅H₄N)CH₂-]n Boiling Pt: 225 °C/ 260 °C (dec. air/ dec. N₂) Density: 1,15 |
MDL Number:
MFCD00190588 CAS Number: 9017-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...